Learn More
Epredia™ Richard-Allan Scientific™ Cover Glass
Shop All Epredia Products
Click to view available options
Thickness (Metric):
#1.5 (0.16 to 0.19 mm)
Length (Metric):
24 mm
Width (Metric):
60 mm
Unit Size:
Each
Description
Ordering Information:
One oz. pack
Specifications
Specifications
| Length (Metric) | 24 mm |
| Width (Metric) | 60 mm |
| Thickness (Metric) | #1.5 (0.16 to 0.19 mm) |
| Dimensions (L x W) | 60 x 24 mm |
| Description | Cvr Glass no.1.5 24x60 10 Oz/cs |