Learn More
Epredia™ Richard-Allan Scientific™ Cover Glass
Product Code. 13202943 Shop All Epredia Products
Change view
Click to view available options
Thickness (Metric):
#1.5 (0.16 to 0.19 mm)
Length (Metric):
24 mm
Width (Metric):
60 mm
Unit Size:
Each
Product Code. Thickness (Metric) Length (Metric) Width (Metric) unitSize
13202943 #1.5 (0.16 to 0.19 mm) 24 mm 60 mm Each
1 options
Product Code. 13202943
Brand: Epredia™ 152460
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Reduces defect costs that range from wasted resources to lost time with a consistent, reliable workflow, which includes quality cover glass.
Ordering Information:
One oz. pack
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 24 mm
Width (Metric) 60 mm
Thickness (Metric) #1.5 (0.16 to 0.19 mm)
Dimensions (L x W) 60 x 24 mm
Description Cvr Glass no.1.5 24x60 10 Oz/cs