missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RIC8A Polyclonal antibody specifically detects RIC8A in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | RIC8A |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | RIC8, RIC8A resistance to inhibitors of cholinesterase 8 homolog A (C. elegans), synembryn, synembryn-A |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 362-531 of human RIC8A (NP_001273063.1).,, Sequence:, DVRTRPEVGEMLRNKLVRLMTHLDTDVKRVAAEFLFVLCSESVPRFIKYTGYGNAAGLLAARGLMAGGRPEGQYSEDEDTDTDEYKEAKASINPVTGRVEEKPPNPMEGMTEEQKEHEAMKLVTMFDKLSRNRVIQPMGMSPRGHLTSLQDAMCETMEQQLSSDPDSDPD |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?