missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RIC3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15761962 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15761962 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15761962 Supplier Invitrogen™ Supplier No. PA564196

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LSKGKTTAED GKCYTAMPGN THRKITSFEL AQLQEKLKET EAAMEKLINR VGPNGERAQT VTSDQEKRLL HQLREITRVM Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.

Promotes functional expression of homomeric alpha-7 and alpha-8 nicotinic acetylcholine receptors at the cell surface. May also promote functional expression of homomeric serotoninergic 5- HT3 receptors, and of heteromeric acetylcholine receptors alpha- 3/beta-2, alpha-3/beta-4, alpha-4/beta-2 and alpha-4/beta-4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIC3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RIC3
Gene Accession No. Q7Z5B4
Gene Alias AYST720; E130307J04Rik; PRO1385; Protein RIC-3; putative alpha7-nAChR chaperone; resistance to inhibitors of cholinesterase 3 homolog; resistance to inhibitors of cholinesterase 3 homolog (C. elegans); resistance to inhibitors of cholinesterase 3-like protein; resistant to inhibitor of cholinesterase 3; Ric3; RIC3 acetylcholine receptor chaperone; UNQ720/PRO1385
Gene Symbols RIC3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RIC3. Recombinant protein control fragment (Product #RP-103581).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79608
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.