missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribosomal protein L4 Rabbit anti-Human, Clone: 3K6, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80959-1-RR-20UL
Additional Details : Weight : 0.00010kg
Description
Ribosomal protein L4 antibody apecifically detetects Ribosomal protein L4 antigen in Human Samples. It is validated for Western Blot, ELISA.Specifications
Ribosomal protein L4 | |
3K6 | |
Unconjugated | |
BC009888 | |
Rabbit | |
Protein A purification | |
Primary | |
Human | |
Liquid |
Western Blot, ELISA | |
1000 μg/mL | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
RPL4 | |
Ribosomal protein L4 fusion protein Ag28728 | |
20 μL | |
6124 | |
Store at -20°C. Stable for one year after shipment. | |
IgG |