Learn More
Invitrogen™ RiboMinus™ Eukaryote Kit v2
Product Code. 15880833
Change view
Click to view available options
Quantity:
12 reactions
Unit Size:
12 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15880833 12 reactions 12 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15880833 Supplier Invitrogen™ Supplier No. A15020
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
The RiboMinus™ Eukaryote Kit v2 improves upon the previous system (RiboMinus™ Eukaryote Kit for RNA-Seq) by removing significantly more rRNA (both cytoplasmic and mitochondrial) from the total RNA.
Features of the kit include:
• Seamless integration with RiboMinus™Concentration Module (available separately) to purify RNA after ribosomal RNA depletion
• 1.5-hour workflow
• Input range of 1-5 μg
The RiboMinus™ Eukaryote Kit v2 improves upon the previous system (RiboMinus™ Eukaryote Kit for RNA-Seq) by removing significantly more rRNA (both cytoplasmic and mitochondrial) from the total RNA. When used with our library construction kits for sequencing, the RiboMinus™ Eukaryote Kit v2 provides maximum mapping to reference database with minimum bias, maintaining relative gene expression levels of even smaller RNA species. The resulting total RNA after depletion contains minimum contaminating rRNA with reduced variability.
The RiboMinus Eukaryote Kit v2 is designed for the removal of human, mouse, and rat rRNA. The RiboMinus™ Concentration Module (available separately) is used for RNA purification after rRNA depletion. For magnetic bead-based purification, please see the RiboMinus™ Eukaryote System v2. The RiboMinus Eukaryote Kit v2 has an RNA input range of 1-5 μg. For lower inputs (down to 100 ng), the Low Input RiboMinus™ Eukaryote System v2 is recommended.
The RiboMinus™ Eukaryote Kit v2 contains sufficient reagents to perform 12 ribosomal RNA depletions.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage
• RiboMinus™ Eukaryote Probe Mix, 2-20°C
• RiboMinus™ Magnetic Beads, 4°C
• 2 x Hybridization Buffer, room temperature
Isolation Technology Magnetic Bead
Species Human, Mouse, Rat
Final Product Type Transcriptome RNA
For Use With (Equipment) Ion PGM™ System, Ion Proton™ System, Illumina™ Genome Analyzer II (GA II)
Product Line RiboMinus
Quantity 12 reactions
Sequencing Type Transcriptome Sequencing
Starting Material Amount 1 - 5 μg total RNA
Workflow Step Sample Enrichment