missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin, MedChemExpress

Product Code. p-200099158
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
16490938 5 g 5g
16412189 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 16490938

Brand: Medchem Express HYB0456

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Riboflavin is an easily absorbed micronutrient with a key role in maintaining health in humans and other animals.

Riboflavin is an easily absorbed micronutrient with a key role in maintaining health in humans and other animals.

Specifications

Chemical Name or Material Riboflavin
Physical Form Solid
CAS 83-88-5
Color Deep Yellow
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C17H20N4O6
Synonym Vitamin B2 E101
Solubility Information DMSO : 17.86 mg/mL (47.45 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 14.29 mg/mL (37.97 mM; ultrasonic and adjust pH to 8 with NaOH)
SMILES O[C@H]([C@H](O)[C@H](O)CO)CN1C(C=C(C)C(C)=C2)=C2N=C(C(N3)=O)C1=NC3=O
Molecular Weight (g/mol) 376.36
Formula Weight 376.36
Percent Purity 98.11%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.