missing translation for 'onlineSavingsMsg'
Learn More

RHPN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7204978 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16870514 20 μL 20µL
16860514 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16870514

Brand: Proteintech 126711AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The MW of this protein is 77 kDa, and this antibody specifically recognizes the 77 kDa protein.

This gene encodes a member of the rhophilin family of Ras-homologous (Rho)-GTPase binding proteins. The encoded protein binds both GTP- and GDP-bound RhoA and GTP-bound RhoB and may be involved in the organization of the actin cytoskeleton.

Specifications

Antigen RHPN2
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RHPN2
Gene Accession No. Q8BWR8, Q8IUC4
Gene Alias 76 kDa RhoB effector protein, GTP Rho binding protein 2, p76RBE, RhoBP, Rhophilin 2, RHPN2
Gene Symbols Rhpn2
Host Species Rabbit
Immunogen RHPN2 Fusion Protein Ag3353
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308516, 52428, 85415
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.