Learn More
Rhodizonic acid dihydrate, 98%
Click to view available options
Quantity:
1 g
Unit Size:
1g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.
Soluble in water.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.
Specifications
Specifications
| CAS | 63183-44-8 |
| Melting Point | ∽248°C (decomposition) |
| Molecular Formula | C6H2O6·2H2O |
| MDL Number | MFCD00149072 |
| Quantity | 1 g |
| Beilstein | 2047007 |
| Synonym | 2-cyclohexene-1,4-dione, 2,3,5,5,6,6-hexahydroxy, hexahydroxycyclohex-2-ene-1,4-dione, 2,2,3,3,5,6-hexahydroxy-5-cyclohexene-1,4-dione, 2-cyclohexene-1,4-dione,2,3,5,5,6,6-hexahydroxy |
| Solubility Information | Soluble in water. |
| Formula Weight | 206.11 (170.09 Anhydrous) |
| Percent Purity | 98% |
| Show More |