Learn More
Rhodium solution for ICP 1g/L HCl matrix, Fisher chemical™
BIOPHARMA
Product Code. 12993484 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
Unit Size:
100mL
250mL
500mL
Product Code. 12993484
Brand: Fisher Chemical S 047 250
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Bottle
Analyte or Component Names Rh
Concentration 1000 g/L
Type Mass Spectrometry Standard
Element Rh
Solvent or Matrix 5% HCl
Quantity 250 mL
compliance-icons
Product Identifier
  • Single-element Standard Solution for ICP. Rhodium (Rh) 1000mg/l in HCl 5% (S 047)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.