Learn More
MedChemExpress Rhodamine 123
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Specifications
Specifications
| Content And Storage | 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Quantity | 25 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : 100 mg/mL (262.59 mM; Need ultrasonic) |
| Target | HBV |
| Product Type | Dye Reagents |
| Molecular Weight (g/mol) | 380.82 g/mol |
| CAS | 62669-70-9 |
| Synonym | RH-123 ∣ R-22420 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.