Description
Thermo Scientific™ RheFlex™ Two-Piece PEEK Fittings provide inert, metal-free connections.
- Slotted back-side of the ferrule is squeezed down onto the tube by the mating conical surface of the nut
- May be used on 1/16 in. metal or plastic tubing reliably up to 5000 psi
- Reusable ferrule and nut
Specifications
Specifications
| Type | Replacement Ferrule |
| Description | Replacement ferrules; 5/Pk. |
| For Use With (Equipment) | PEEK Tubing |
| Material | PEEK |