Description
Ensure reliable high-pressure connections by using Thermo Scientific™ RheFlex™ High-Pressure Fittings. These fittings are precision machined from 316 stainless steel to be burr- and contaminant-free.
Specifications
Specifications
| Type | Long High Pressure Fittings Set |
| Description | 10-32, 1/16 in. Long Fittings Set |
| For Use With (Equipment) | LC Columns |
| Material | Stainless Steel |
| Quantity | 5/Pk. |