Learn More
Thermo Scientific™ RheFlex™ High-Pressure Fittings
Product Code. 10016844 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Extra Long High Pressure Fittings Set
High Pressure 1/16 in. Ferrule
Long High Pressure Fittings Set
Short High Pressure Fittings Set
Quantity:
10/Pk.
5/Pk.
Unit Size:
Pack of 10
Pack of 5
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Quantity unitSize
10016844 Short High Pressure Fittings Set 5/Pk. Pack of 5
10650816 High Pressure 1/16 in. Ferrule 5/Pk. Pack of 5
10249704 Long High Pressure Fittings Set 5/Pk. Pack of 5
11567701 Extra Long High Pressure Fittings Set 5/Pk. Pack of 5
11577701 Short High Pressure Fittings Set 10/Pk. Pack of 10
10299594 High Pressure 1/16 in. Ferrule 10/Pk. Pack of 10
10269264 Long High Pressure Fittings Set 10/Pk. Pack of 10
10197704 Extra Long High Pressure Fittings Set 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10016844 Supplier Thermo Scientific™ Supplier No. 6000109
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ensure reliable high-pressure connections with Thermo Scientific™ RheFlex™ High-Pressure Fittings.
Ensure reliable high-pressure connections by using Thermo Scientific™ RheFlex™ High-Pressure Fittings. These fittings are precision machined from 316 stainless steel to be burr- and contaminant-free.
TRUSTED_SUSTAINABILITY
Specifications
Type Short High Pressure Fittings Set
Description SS Standard Fittings; 1/16 in.
For Use With (Equipment) LC Columns
Material Stainless Steel
Quantity 5/Pk.