missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RHEB Recombinant Protein

Brand:  Abnova™ H00006009-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16190222

  • 6245.00 NOK / 25µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH16155
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
45.98
8
Glutathione Sepharose 4 Fast Flow
25 ug
MPQSKSRKIAILGYRSVGKSSLTIQFVEGQFVDSYDPTIENTFTKLITVNGQEYHLQLVDTAGQDEYSIFPQTYSIDINGYILVYSVTSIKSFEVIKVIHGKLLDMVGKVQIPIMLVGNKKDLHMERVISYEEGKALAESWNAAFLESSAKENQTAVDVFRRIILEAEKMDGAASQGKSSCSVM
MGC111559/RHEB2
RHEB
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
6009
RHEB (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RHEB
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers