Learn More
RHEB Mouse anti-Human, Mouse, Clone: 2F9H8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66714-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
Rheb encodes a member of the small GTPase superfamily and encodes a lipid-anchored, cell membrane protein with five repeats of the RAS-related GTP-binding region. This protein is vital in regulation of growth and cell cycle progression due to its role in the insulin/TOR/S6K signaling pathway. The protein has GTPase activity and shuttles between a GDP-bound form and a GTP-bound form, and farnesylation of the protein is required for this activity. Three pseudogenes have been mapped, two on chromosome 10 and one on chromosome 22.Specifications
RHEB | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15382, Q921J2 | |
RHEB | |
RHEB Fusion Protein Ag8887 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Western Blot | |
2F9H8 | |
Unconjugated | |
RHEB | |
GTP binding protein Rheb, Ras homolog enriched in brain, RHEB, RHEB2 | |
Mouse | |
Protein G | |
RUO | |
19744, 6009 | |
-20°C | |
Liquid |