missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11590-1-AP-20UL
This item is not returnable.
View return policy
Description
The 42 kDa band recognized by this antibody is a RGS5-RGS5 dimer.
The regulator of G protein signaling (RGS) proteins are signal transduction molecules that have structural homology to SST2 of Saccharomyces cerevisiae and EGL-10 of Caenorhabditis elegans. Multiple genes homologous to SST2 are present in higher eukaryotes. RGS proteins are involved in the regulation of heterotrimeric G proteins by acting as GTPase activators. The mRNA of the human gene is abundantly expressed in heart, lung, skeletal muscle, and small intestine, and at low levels in brain, placenta, liver, colon, and leukocytes.Specifications
| RGS5 | |
| Polyclonal | |
| Unconjugated | |
| RGS5 | |
| MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129, RGS5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19737, 54294, 8490 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08850, O15539, P49800 | |
| Rgs5 | |
| RGS5 Fusion Protein Ag2196 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction