Learn More
RGS14 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16258-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the regulator of G-protein signaling family. This protein contains one RGS domain, two Raf-like Ras-binding domains (RBDs), and one GoLoco domain. The protein attenuates the signaling activity of G-proteins by binding, through its GoLoco domain, to specific types of activated, GTP-bound G alpha subunits. Acting as a GTPase activating protein (GAP), the protein increases the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.Specifications
| RGS14 | |
| Polyclonal | |
| Unconjugated | |
| RGS14 | |
| RGS14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10636, 51791 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43566, P97492 | |
| Rgs14 | |
| RGS14 Fusion Protein Ag9292 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |