missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RGS1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16363595
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16363595 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16363595 Supplier Invitrogen™ Supplier No. PA586730

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

This gene encodes a member of the regulator of G-protein signaling family. This protein is located on the cytosolic side of the plasma membrane and contains a conserved, 120 amino acid motif called the RGS domain. The protein attenuates the signaling activity of G-proteins by binding to activated, GTP-bound G alpha subunits and acting as a GTPase activating protein (GAP), increasing the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RGS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene RGS1
Gene Accession No. P97844, Q08116, Q9JL25
Gene Alias 1R20; B-cell activation protein BL34; BL34; Early response protein 1R20; epididymis secretory protein Li 87; HEL-S-87; IER1; immediate-early response 1, B-cell specific; IR20; Regulator of G-protein signaling 1; regulator of G-protein signalling 1; RGS1
Gene Symbols RGS1
Host Species Rabbit
Immunogen Synthetic peptide, corresponding to amino acids 161-206 of Human RGS1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50778, 54289, 5996
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.