Learn More
MedChemExpress RGD Trifluoroacetate
Specifications
Specifications
| Purity | 98.0% |
| CAS | 2378808-45-6 |
| Content And Storage | Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) |
| For Use With (Application) | Cancer-programmed cell death |
| Molecular Weight (g/mol) | 460.36 g/mol |
| Product Type | Peptides |
| Quantity | 100 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : ≥ 60 mg/mL (130.33 mM) |