missing translation for 'onlineSavingsMsg'
Learn More

RG7800 hydrochloride, MedChemExpress

Product Code. 16466297
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16466297 5 mg 5mg
16439986 50 mg 50mg
16424686 100 mg 100mg
16423578 10 mg 10mg
4 options
This item is not returnable. View return policy

Product Code. 16466297

Brand: Medchem Express HY101792A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RG7800 hydrochloride is an orally active SMN2 splicing modulator, with EC1.5xs of 23 nM and 87 nM for SMN2 splicing and SMN protein; RG7800 hydrochloride has the potential to treat spinal muscular atrophy.

RG7800 hydrochloride is an orally active SMN2 splicing modulator, with EC1.5xs of 23 nM and 87 nM for SMN2 splicing and SMN protein

Specifications

Chemical Name or Material RG7800 hydrochloride
Physical Form Solid
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C24H28N6O.xHCl
For Use With (Application) Neuroscience-Neuromodulation
Synonym RO6885247 hydrochloride
Solubility Information H2O : ≥ 113.5 mg/mL
SMILES CC1=CC(C2CCN(C)CC2)=CN3C1=NC(C4=NN(C=C(C)N=C5CC)C5=C4)=CC3=O.[xHCl]
Percent Purity 99.59%
Grade Research
Visa mer Visa mindre

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.