missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFX1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26859-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the regulatory factor X gene family, which encodes transcription factors that contain a highly-conserved winged helix DNA binding domain. The protein encoded by this gene is structurally related to regulatory factors X2, X3, X4, and X5. It is a transcriptional activator that can bind DNA as a monomer or as a heterodimer with RFX family members X2, X3, and X5, but not with X4. This protein binds to the X-boxes of MHC class II genes and is essential for their expression. Also, it can bind to an inverted repeat that is required for expression of hepatitis B virus genes.Specifications
RFX1 | |
Polyclonal | |
Unconjugated | |
RFX1 | |
EF C, Enhancer factor C, Regulatory factor X 1, RFX, RFX1, Transcription factor RFX1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5989 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P22670 | |
RFX1 | |
RFX1 Fusion Protein Ag25442 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |