Learn More
Thermo Scientific™ Reverse Osmosis Membranes
Product Code. 11377131 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Reverse Osmosis Membrane
Reverse Osmosis Membrane (pretreated), 3 L/hr.
Reverse Osmosis Membrane (pretreated), 6 L/hr.
For Use With (Equipment):
LabTower EDI 15, LabTower RO 20/60, LabTower TII 20/60, Pacific RO 3/7/12/20, Pacific TII 3/7/12/20
LabTower EDI 30, LabTower RO 40/60, LabTower TII 40/60, Pacific RO 40, Pacific TII 40
ROpure Infinity Systems
Smart2Pure
Unit Size:
Each
Product Code. Type For Use With (Equipment) unitSize
11377131 Reverse Osmosis Membrane (pretreated), 3 L/hr. Smart2Pure Each
11377161 Reverse Osmosis Membrane LabTower EDI 30, LabTower RO 40/60, LabTower TII 40/60, Pacific RO 40, Pacific TII 40 Each
11830820 Reverse Osmosis Membrane ROpure Infinity Systems Each
11327141 Reverse Osmosis Membrane (pretreated), 6 L/hr. Smart2Pure Each
11337161 Reverse Osmosis Membrane LabTower EDI 15, LabTower RO 20/60, LabTower TII 20/60, Pacific RO 3/7/12/20, Pacific TII 3/7/12/20 Each
5 options
Product Code. 11377131
Brand: Thermo Scientific™ 09.2003
Only null left
Add to basket
Add to basket
Choose the required Thermo Scientific™ Reverse Osmosis (RO) Membrane for your Thermo Scientific™ Barnstead™ point of use water purification system, as indicated.
Use Membranes with the Following Barnstead Systems:
09.2003:
  • Reverse osmosis membrane with integrated pretreatment, 3L/hr for Smart2Pure™ systems
09.2006:
  • Reverse osmosis membrane with integrated pretreatment, 6L/hr for Smart2Pure systems
22.0046:
  • LabTower™ EDI 15 requires two of these RO membranes
  • LabTower™ RO 20 requires two of these RO membranes
  • LabTower RO 60 requires two of these RO membranes, along with two 22.0087 RO membranes
  • LabTower™ TII 20 requires two of these RO membranes
  • LabTower TII 60 requires two of these RO membranes, along with two 22.0087 RO membranes
  • Pacific™ RO 3, 7, 12 require one of these RO membranes
  • Pacific RO 20 requires two of these RO membranes
  • Pacific™ TII 3, 7, 12 require one of these RO membranes
  • Pacific TII 20 requires two of these RO membranes
22.0087:
  • LabTower™ EDI 30 requires two of these RO membranes
  • LabTower™ RO 40 requires two of these RO membranes
  • LabTower RO 60 requires two of these RO membranes, along with two 22.0046 RO membranes
  • LabTower TII 40 requires two of these RO membranes
  • LabTower TII 60 requires two of these RO membranes, along with two 22.0046 RO membranes
  • Pacific RO 40 requires two of these RO membranes
  • Pacific TII 40 requires two of these RO membranes
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Smart2Pure
Type Reverse Osmosis Membrane (pretreated), 3 L/hr.