Learn More
Masterflex™ Reusable Acetal Copolymer Hose Clamp Kit
Click to view available options
Color:
White
Unit Size:
Each
Description
Recommended for continuous use from ambient temperature to 93°C. Clamps are non-conducting and stable in a wide variety of chemicals. Once set, clamps stay put. To use, just position clamp on hose or tubing bundle and press the jaws together with a pair of pliers. White Acetal Hose Clamps Kit includes 170 assorted clamps from 0.228 to 1.405 in. ID.
Specifications
Specifications
| Material | Acetal Copolymer |
| Packaging Quantity | 1 Pack |
| Color | White |
| Product Type | Hose Clamp Kit |