Learn More
Masterflex™ Reusable White Acetal Copolymer Hose Clamp
Product Code. 12363830
Change view
Click to view available options
For Use With:
0.23 x 0.26 in. Tubing I.D.
0.35 x 0.39 in. Tubing I.D.
0.40 x 0.45 in. Tubing I.D.
0.51 x 0.59 in. Tubing I.D.
0.61 x 0.70 in. Tubing I.D.
Unit Size:
Pack of 100
Product Code. For Use With unitSize
12363830 0.23 x 0.26 in. Tubing I.D. Pack of 100
11755169 0.61 x 0.70 in. Tubing I.D. Pack of 100
12409660 0.35 x 0.39 in. Tubing I.D. Pack of 100
15335922 0.40 x 0.45 in. Tubing I.D. Pack of 100
12429660 0.51 x 0.59 in. Tubing I.D. Pack of 100
5 options
Product Code. 12363830
Brand: Masterflex™ 0683201
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Material Acetal Copolymer
For Use With 0.23 x 0.26 in. Tubing I.D.
Packaging Quantity 100 Pack
Color White
Product Type Hose Clamp