Learn More
Masterflex™ Reusable White Acetal Copolymer Hose Clamp
Specifications
Specifications
| Material | Acetal Copolymer |
| For Use With | 0.61 x 0.70 in. Tubing I.D. |
| Packaging Quantity | 100 Pack |
| Color | White |
| Product Type | Hose Clamp |