Learn More
3M™ Reusable Half Face Mask and A2P3 Combination Filter Kit
This handy, ready-to-use kit includes a 3M™ Reusable Half Face Mask and an A2P3 filter, all in a reusable storage box.
Brand: 3M™ 7100265072
6410.00 NOK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 5.18300kg
Description
- Includes 1 x 3M™ Reusable Half Face Mask and 1 x 3M™ Combination Filter CF32 A2P3 R D DT-4041E
- Offers A2P3 protection from certain organic vapors and particulate hazards
- Comes in a useful, reusable storage box
Specifications
Ready-to-Use Respirator Pack | |
3M™ Reusable Half Face Mask and an A2P3 Filter | |
10/Cs. |
Small | |
HF-3014 |