Learn More
Retsch Retsch Test sieve 200x50mm 450µm ISO 3310/1
Brand:  Retsch 60.131.000450
Retsch Test sieve 200x50mm 450µm ISO 3310/1
Product Code. 15650279
  • ONLINE EXCLUSIVE PRICE
    1740.00 NOK
    1616.52 NOK / Each
    Save 123.48 NOK
    7% Off
Only null left
Estimated Shipment: 20-04-2026