missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Resurge™ CD5 Supplement, without glucose and L-glutamine
Click to view available options
Quantity:
100 g
1 kg
5 kg
Unit Size:
100g
1kg
5kg
Description
A chemically defined supplement formulation to complement a variety of base media. This supplement was developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. Developed for use in CHO mammalian culture systems and can be used either as a media supplement or a culture system feed.
Specifications
Specifications
| Content And Storage | Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers. |
| Cell Line | CHO |
| Classification | Animal Origin-free, Chemically-defined, Protein-free |
| Culture Type | Mammalian Cell Culture |
| Form | Powder |
| Product Type | Media Supplement |
| Without Additives | No Glutamine |
| Format | Bottle(s) |
| Product Line | Resurge |
| Quantity | 1 kg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction