Learn More
Gibco™ Resurge™ CD3 Supplement, without glucose and L-glutamine
Description
An animal origin-free (AOF) protein-free chemically defined supplement. It can be used in batch and fed batch mode in cell culture processes. All components are chemically defined with known molecular formulae and structures and meet our stringent Animal Origin-Free (AOF) specification. Formulated without glucose, L-glutamine, and sodium bicarbonate. Due to the balanced nutritional nature of this supplement, rich in amino acids, it has also been used in bacterial applications.
Specifications
Specifications
| Content And Storage | Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers. |
| Cell Line | CHO |
| Classification | Animal Origin-free, Chemically-defined, Protein-free |
| Form | Powder |
| Product Type | Media Supplement |
| Without Additives | No Glutamine, No Glucose |
| Format | Bottle(s) |
| Product Line | Resurge |
| Quantity | 100 g |
| Source | Animal origin-free |