Learn More
Velp Scientifica™ RESPIROMETRIC Sensor System 6 for Soil Analysis
Product Code. 17193174 Shop All Velp Products
Change view
Click to view available options
Includes:
RESPIROMETRIC Sensor (x6), Kit for respirometric analysis of soil (x6)
RESPIROMETRIC Sensor (x6), RESPIROSoft and Wireless DataBox, Kit for respirometric analysis of soil (x6)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
17193174 RESPIROMETRIC Sensor (x6), Kit for respirometric analysis of soil (x6) Each
17163174 RESPIROMETRIC Sensor (x6), RESPIROSoft and Wireless DataBox, Kit for respirometric analysis of soil (x6) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17193174 Supplier Velp Scientifica™ Supplier No. SB102B0176
  • ONLINE EXCLUSIVE PRICE
    33940.00 NOK
    30600.61 NOK / Pack of 1
    Save 3339.39 NOK
    9% Off
Estimated Shipment: 05-01-2027
The versatile, reliable and cloud-enabled solution designed for aerobic degradation tests on soil and compost.
  • Designed to support aerobic degradation tests on soil and compost. The analysis provides microbial soil respiration results from the mineralization of organic substances. A major application of these tests is related to the detection and monitoring processes during decontamination and clean-ups.
  • The bright 4-digit LED display ensures long battery life in respect of other sensors in the market.
  • Intuitive 2 button interface to facilitate the analysis set up
  • Pressure range: 500 ÷ 2000 mbar (hPa)
  • Results available in mbar
  • The RESPIROMETRIC Sensor transmits data directly to the PC enabling real-time monitoring of the analysis curve. Respirometric studies can last days and even months, the unique VELP solution ensures constant monitoring of multiple analysis from an intuitive proprietary software, the RESPIROSoft™
  • Connect the RESPIROMETRIC Sensor to the exclusive ELP Ermes Cloud Platform to improve your laboratory experience
    The VELP Ermes Cloud Platform connection will unburden you from tedious tasks improving your lab productivity
  • Includes: RESPIROMETRIC Sensor ( x 6), MULTISTIRRER6 Digital 100-240 V/50-60 Hz (with bottles and stirring bars), Kit for respirometric analysis of plastics ( x 6)
Specifications
Display Type Digital
Battery Type CR 2430
Connection Type Ermes via Wi-Fi / RESPIROSoft™ via USB
Material Technopolymer
Range 500/2000 Mbar (hPa)
Volume (Metric) Sample Bottle volume: 1000 mL
For Use With (Application) Aerobic Degradation Tests on Soil and Compost.
Includes RESPIROMETRIC Sensor (x6), Kit for respirometric analysis of soil (x6)
Warranty 25 months
Weight (Metric) 1.5 kg