Description
- Preformed mask
- Labelled for use with high levels of dust exposure (D)
Specifications
Specifications
| Certifications/Compliance | EN 149 |
| Color | White |
| Design Type | Flat-fold |
| Disposable | Single-use |
| Filter Type | FFP3 |
| Material | Polypropylene, Polyester |
| Product Type | Particulate Respirator |
| Size | Regular |
| With Exhalation Valve | Yes |
| Model | silv-Air c 3310 FFP3 |