missing translation for 'onlineSavingsMsg'
Learn More

Resorcinol, MP Biomedicals™

Product Code. 11426192 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 11426192

Brand: MP Biomedicals 0210280890

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Resorcinol is an aromatic alcohol used in large scale processes, dermatology research, and in colorimetric assays for sialic acid.

  • Resorcinol is an aromatic alcohol used in large scale processes such as tanning, the manufacture of resins and resin adhesives, the production of hexylresorcinol and p-aminosalicylic acid, and the dyeing and printing of textiles.
  • Resorcinol is used in dermatology research, such as in studies of irritant contact dermatitis.
  • The role of resorcinol in causing cytotoxicity and cytokine release in human epidermal keratinocytes and dermal fibroblasts has been studied.
  • The cytoxicity of resorcinol and other phenolic compounds against the melanotic human melanoma cell lines IRE 1 and IRE 2, and the lymphoma- and leukemia-derived cell lines Raji and K 562 has been studied, at phenolic compound concentrations of 10 μM to 5 mM.
  • The use of resorcinol in a colorimetric assay for sialic acid has been reported.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-46-3
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00002269
InChI Key GHMLBKRAJCXXBS-UHFFFAOYSA-N
Synonym resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
PubChem CID 5054
ChEBI CHEBI:27810
IUPAC Name benzene-1,3-diol
SMILES C1=CC(=CC(=C1)O)O

Specifications

Melting Point 228 to 232°F
Color White
Density 1.2 g/cm3 at 68°F
Boiling Point 531°F to 536°F (at 760 mm Hg)
Flash Point 261°F
Ignition Point 1125°F
Assay Percent Range ≥99%
Quantity 500 g
Solubility Information ≥100 mg/mL at 65.3°F
Vapor Density 3.79
Formula Weight 110.11
Vapor Pressure 1 mm Hg at 227.1°F, 0.0002 mm Hg at 77°F
Percent Purity ≥99%
Physical Form Solid
For Use With (Application) Building Blocks, Chemical Synthesis
Chemical Name or Material Resorcinol, Molecular Biology, Cell Biology
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.