Learn More
WTW™ Resistance Test Kit for WTW™ Conductivity Meter
Product Code. 10191721
Change view
Click to view available options
For Use With (Equipment):
All Conductivity Meters
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10191721 All Conductivity Meters Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10191721 Supplier WTW™ Supplier No. 302870
 Restricted Item
In Stock
For conductivity measuring systems, there are different check methods:The system including measuring cell can be calibrated with suitable traceably certifed standard solutions. An adjustment is usually not necessary due to the long-term stability of the used electrode materials. Special kits contain all necessary parts to test conductivity measuring systems according to pharamcopeia guidelines.
TRUSTED_SUSTAINABILITY
Specifications
Description Resistance test kit
Includes Waterproof 8-pin socket
For Use With (Equipment) All Conductivity Meters