Learn More
Thermo Scientific™ Reservoir Accessories
Product Code. 11347171 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Sterile Vent Filter
Vent Filter/CO2 Adsorber and Sterile Overflow
For Use With (Equipment):
Reservoir, 1100 to 2000 L
Storage Tank
Storage Tank 30 to 100 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Unit Size
11347171 Sterile Vent Filter Storage Tank Each
15172296 Sterile Vent Filter Storage Tank 30 to 100 L Each
15514780 Vent Filter/CO2 Adsorber and Sterile Overflow Reservoir, 1100 to 2000 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11347171 Supplier Thermo Scientific™ Supplier No. 22.0091
  • ONLINE EXCLUSIVE PRICE
    1540.00 NOK
    1450.65 NOK / Pack of 1
    Save 89.35 NOK
    5% Off
Estimated Shipment: 17-11-2026
Level control and sterile overflow for storage reservoirs are perfect accessories for running reverse osmosis systems.
Level control and sterile overflow, Thermo Scientific™ Reservoir Accessories, are perfect for running reverse osmosis systems.
Level Control for Storage Reservoirs
  • Required accessory for running any reverse osmosis system; will be adapted to the specific reservoir during the order procedure
Sterile Overflow for Storage Reservoirs
  • Recommended accessory to prevent organic contamination inside of reverse osmosis reservoirs
Vent filter for Storage Reservoirs
  • Recommended accessory to prevent organic contamination inside of reverse osmosis reservoirs
Vent filter with CO2 absorber for Storage Reservoirs
  • Recommended accessory to prevent organic and CO2 contamination inside of reverse osmosis reservoirs
Ordering Information:
This product is not available in all regions
TRUSTED_SUSTAINABILITY
Specifications
Description 0.2 μm
For Use With (Equipment) Storage Tank
Type Sterile Vent Filter