Learn More
Gosselin™ Resealable Plastic Bag, Double Pocket, PS
Product Code. 15497451
Click to view available options
Dimensions (L x W):
200mm x 150mm
Unit Size:
Pack of 4000
Product Code. 15497451
Brand: Gosselin™ SP438
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Corning™ Gosselin™ Resealable Plastic Bag, Double Pocket, PS, are suitable for a variety of storage uses.
  • Clear, polystyrene construction
  • Resealable
  • Double pocket design
TRUSTED_SUSTAINABILITY
Specifications
Material Polystyrene
No. per Case 4000/Cs
Quantity 4000/Cs.
Dimensions (L x W) 200mm x 150mm
Width (Metric) 150 mm
Height (Metric) 200 mm