missing translation for 'onlineSavingsMsg'
Learn More

RERE Polyclonal Antibody, Invitrogen™

Product Code. 15752901 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15752901 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15752901 Supplier Invitrogen Supplier No. PA554975

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MCDGGSTEDG CVAASRDDTT LNALNTLHES GYDAGKALQR LVKKPVPKLI EKCWTEDEVK RFVKGLRQYG KNFFRIRKEL LPNKETGELI TFYYYWKKTP EAASSRAHRR HRRQAVFRRI KTRTASTPVN TPSR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

RERE encodes a member of the atrophin family of arginine-glutamic acid (RE) dipeptide repeat-containing proteins. The encoded protein co-localizes with a transcription factor in the nucleus, and its overexpression triggers apoptosis. A similar protein in mouse associates with histone deacetylase and is thought to function as a transcriptional co-repressor during embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RERE
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RERE
Gene Accession No. Q9P2R6, Q80TZ9, Q62901
Gene Alias ARG, ARP, ATN1L, DNB1, arginine-glutamic acid dipeptide repeats protein, atrophin 2, atrophin-1 like protein, atrophin-1 related protein, atrophin-1-like protein, atrophin-1-related protein
Gene Symbols RERE
Host Species Rabbit
Immunogen Recombit protein corresponding to Human RERE
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116665, 473, 68703
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.