Learn More
Cole-Parmer™ Replacement UV Tubes
Product Code. 11830250 Shop All Cole-Parmer Products
Change view
Click to view available options
Wavelength:
365 nm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength Unit Size
11830250 365 nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11830250 Supplier Cole-Parmer™ Supplier No. 0981555
  • ONLINE EXCLUSIVE PRICE
    1155.00 NOK
    1064.26 NOK / Pack of 1
    Save 90.74 NOK
    7% Off
Estimated Shipment: 26-01-2027
Use these replacement UV tubes with ultraviolet lamps.
  • Replacement Ultraviolet Tubes
  • Product may contain trace amounts of mercury
  • Use caution with breakage, cleanup, shipping and/or disposal
Specifications
Wattage 15 W
Wavelength 365 nm
Diameter (Metric) 25.5 mm
Product Type UV Tube