Learn More
BRAND™ seripettor™ / seripettor™ pro Pump Assembly
Shop All Brand Products
Click to view available options
For Use With (Equipment):
Seripettor pro, 25 mL
Seripettor, 10 mL
Seripettor, 25 mL
seripettor, 2 mL
Unit Size:
Each
Description
- seripettor™ pump assembly provided with stainless steel lifting spring.
- seripettor™ pro pump assembly provided with Hastaloy™ (stainless) lifting spring, with PEI for UV protection.
Specifications
Specifications
| Product Type | Pump Assembly |
| Autoclavable | Not Autoclavable |
| Color | Dark Blue, Red, Translucent Amber |
| For Use With (Application) | Dispensing liquid |
| For Use With (Equipment) | Seripettor pro, 25 mL |
| Includes | Pump assembly only |
| Sterility | Non-sterile |