Learn More
Elgas™ Replacement Resin Pack
Product Code. 12804072
Change view
Click to view available options
Capacity:
3 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
12804072 3 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12804072 Supplier Elgas™ Supplier No. 761014
  • ONLINE EXCLUSIVE PRICE
    2299.50 NOK
    1609.61 NOK / Pack of 1
    Save 689.89 NOK
    30% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Replacement resin packs for Ioniser Mark 3C and Mark 6C deioniser
Replacement resin packs for Ioniser Mark 3C and Mark 6C deioniser. One pack is used for Mark 3C. Two packs are used for Mark 6C.
Specifications
Capacity 3 L