Description
Thermo Scientific™ Replacement Plungers for Syringes are available for selected syringes from a variety of manufacturers.
Specifications
Specifications
| Description | For use with 365K3041 or 365K3051 |
| Needle Gauge | 22 to 23 G |
| Length (English) Needle | 1.9685 in. |
| Product Type | Replacement Syringe Plunger |
| Volume (Metric) | 1 mL |
| For Use With (Equipment) | Thermo 365K3041 or 365K3051 Syringes |