Learn More
Scientific Industries SI™ Rubber Cover
Product Code. 15507335 Shop All Ohaus Products
Change view
Click to view available options
For Use With (Equipment):
Vortex-Genie 1, Vortex-Genie 2, Vortex-Genie Pulse
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15507335 Vortex-Genie 1, Vortex-Genie 2, Vortex-Genie Pulse Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15507335 Supplier Scientific Industries SI™ Supplier No. 83041442
  • ONLINE EXCLUSIVE PRICE
    558.48 NOK
    374.83 NOK / Pack of 1
    Save 183.65 NOK
    32% Off
View all promotions
Estimated Shipment: 26-11-2026
Fits on 76.2mm Replacement Platform. Scientific Industries SI™ Rubber Cover is used with Vortex-Geinie 1 and Vortex-Genie 2 family of mixers and Vortex-Genie Pulse.
  • For use with Vortex-Genie family of products
  • Platform attachment required
TRUSTED_SUSTAINABILITY
Specifications
Material Rubber
For Use With (Equipment) Vortex-Genie 1, Vortex-Genie 2, Vortex-Genie Pulse
Requires Replacement Platform 15547315