Learn More
Cytiva Replacement Parts for SE 280 Mini-Format Vertical Electrophoresis Units
Brand:  Cytiva SE282N-2
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10055894
  • 1014.84 NOK / Pack of 2
Estimated Shipment: 17-07-2024
Specifications
SE 280 Mini Format Vertical Gel Electrophoresis Unit
Alumina
12 cm
Notched Alumina Plate
10 cm