Learn More
Replacement Parts for Modular burette
For use with modular burette.
Brand: MBL BRB274P
277.40 NOK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.17000kg
Description
- For use with modular burette
Specifications
25 mL | |
Modular Burette | |
Glass | |
Buret Tube |