Learn More
Thermo Scientific™ Replacement Needles for GC Syringes
Product Code. 10351405 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Thermo 365H2301/-F193/-G216/-H238/-I2611/-F1901/-G2091/-J2881 Syringes
Thermo 365K3041 Syringe
Thermo Scientific 36500490 Syringe
Thermo Scientific 36500495, 36503015 Syringes
Thermo Scientific 36500510 Syringe
Thermo Scientific 36500520 Syringe
Thermo Scientific 36550040 Syringe
Thermo Scientific 365D1171 Syringe
Thermo Scientific 365D1841 Syringe
Thermo Scientific 365D3731 Syringe
Length (Metric) Needle:
50 mm
75 mm
80 mm
Unit Size:
Pack of 2
Pack of 5
Product Code. 10351405
Brand: Thermo Scientific™ 365RN225
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
TRUSTED_SUSTAINABILITY
Specifications
Description 365RN225
Needle Gauge 25 G
Product Type Replacement Syringe Needle
Volume (Metric) 500 μL
Length (Metric) Needle 50 mm
Needle Point Style Bevel
For Use With (Equipment) Thermo 365H2301/-F193/-G216/-H238/-I2611/-F1901/-G2091/-J2881 Syringes
Connection Type Removable