Description
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
Specifications
Specifications
| Description | 36566485 |
| Needle Gauge | 23 G |
| Product Type | Replacement Syringe Needle |
| Volume (Metric) | 50 μL |
| Length (Metric) Needle | 50 mm |
| Needle Point Style | Cone |
| For Use With (Equipment) | Thermo Scientific 36500495, 36503015 Syringes |
| Connection Type | Removable |