Learn More
Trajan Scientific and Medical™ Replacement Needles for Agilent Syringes
Click to view available options
Volume (Metric):
50 μL
Unit Size:
Pack of 2
Description
- Needle length is the exposed length of needle
- Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Specifications
| Needle Gauge | 23 G |
| Diameter (Metric) Inner Needle | 0.24 mm |
| Diameter (Metric) Outer Needle | 0.63 mm |
| Volume (Metric) | 50 μL |
| Needle Point Style | Cone |
| For Use With (Equipment) | Agilent Syringes |