Learn More
Trajan Scientific and Medical™ Replacement Needles for Agilent Plunger-in-Needle Syringes
Quality needles to fit a SGE syringe in a range of lengths and tip styles.
Brand:  Trajan Scientific and Medical™ 033730
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15208583
  • 501.00 NOK / Each
Estimated Shipment: 15-07-2024
Description
  • Needle length is the exposed length of needle
  • Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Specifications
Autosampler Syringe Plunger-in-Needle
0.63/0.47 mm
42 mm
23 to 26 G
0.5 μL
Cone