Learn More
SGE™ Replacement Needles for 5μL Syringes
Product Code. 10633903
Change view
Click to view available options
Needle Gauge:
23 G
26 G
Needle Point Style:
Bevel
Cone
Length (English) Needle:
1.9 in.
2.7 in.
Unit Size:
1 set
Each
Pack of 5
Product Code. Needle Gauge Needle Point Style Length (English) Needle unitSize
10633903 23 G Bevel 1.9 in. 1 set
10492731 26 G Bevel 1.9 in. Pack of 5
10065630 23 G Cone 2.7 in. Each
10197350 23 G Cone 1.9 in. Each
4 options
Made of Stainless steel
Product Code. 10633903
Brand: SGE™ 035056
Made of Stainless steel
Made of Stainless steel.
Made of Stainless steel
Specifications
Needle Gauge 23 G
Diameter (English) Inner 0.014 in.
Diameter (English) Outer 0.024 in.
Diameter (Metric) Inner 0.37 mm
Diameter (Metric) Outer 0.63 mm
Length (English) Needle 1.9 in.
Product Type Replacement Needle
Needle Point Style Bevel
For Use With (Equipment) 8 mm O.D. Barrel 5μL Microvolume Syringes