Learn More
Trajan Scientific and Medical™ Replacement Needle and Plunger Kit for Shimadzu Autosampler Syringes
Click to view available options
Needle Gauge:
23 G
26 G
Diameter (Metric) Outer:
0.47 mm
0.63 mm
Unit Size:
Each
Description
For use with Shimadzu Removable Needle Syringes for AOC14, AOC17 and AOC 20 Autosamplers
- Includes spare needle and plunger
- Plunger extends to needle tip
- Sample contained in needle only
- Zero dead volume
- Needle and plunger must be replaced together
Specifications
Specifications
| Needle Gauge | 23 G |
| Diameter (Metric) Outer | 0.63 mm |
| Product Type | Replacement Needle Kit |
| Needle Point Style | Cone |
| For Use With (Equipment) | Syringes (0.5μL) |